Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-(2-Biphenylyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

B2897-1G B2897-5G
TCB2897-1GEA 190 USD InStock
TCB2897-1G TCB2897-5G
9-(2-Biphenylyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
9-(2-Biphenylyl)-10-methylacridinium perchlorate
Molecular Weight: 445.90
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Molecular Formula: C26H20ClNO4


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bfa33f7bed06f6e6ec1eb43847703fb2ffa82ae&vid=1bfa33f7bed06f6e6ec1eb43847703fb2ffa82ae&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty