Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxybenzonitrile ≥98.0%

Supplier: Decon Laboratories Ltd

B3123-5G B3123-25G
TCB3123-5GEA 125.72 USD InStock
TCB3123-5G TCB3123-25G
5-Bromo-2-methoxybenzonitrile ≥98.0%
5-Bromo-2-methoxybenzonitrile
CAS Number: 144649-99-0
MDL Number: MFCD00143429
Molecular Formula: C8H6BrNO
Molecular Weight: 212.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92

Other Product Names / Synonyms: 4-Bromo-2-cyanoanisole


Formula: C₈H₆BrNO
Melting Pt: 92 °C
MDL Number: MFCD00143429
CAS Number: 144649-99-0
UN: 3439
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cf684d9af387c5ec3df98af7d3b2fb4f9e500cc&vid=0cf684d9af387c5ec3df98af7d3b2fb4f9e500cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty