Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Baclofen ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B3343-25G B3343-5G
TCB3343-25GEA 289 USD InStock
TCB3343-25G TCB3343-5G
Baclofen ≥98.0% (by titrimetric analysis)
Baclofen
CAS Number: 1134-47-0
MDL Number: MFCD00055143
Molecular Formula: C10H12ClNO2
Molecular Weight: 213.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 208

Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 4-Amino-3-(4-chlorophenyl)butyric Acid


Formula: C₁₀H₁₂ClNO₂
Melting Pt: 208 °C
MDL Number: MFCD00055143
CAS Number: 1134-47-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a41c77d871501a8537ae08687ebcb6e6051ca7f&vid=4a41c77d871501a8537ae08687ebcb6e6051ca7f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty