Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

B3918-1G B3918-5G
TCB3918-1GEA 74 USD InStock
TCB3918-1G TCB3918-5G
2-Bromo-4-nitrophenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-nitrophenol
CAS Number: 5847-59-6
MDL Number: MFCD06656567
Molecular Formula: C6H4BrNO3
Molecular Weight: 218.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 112
Lambda max.: 404 nm (NaOH)

Other Product Names / Synonyms: 3-Bromo-4-hydroxynitrobenzene


Formula: C₆H₄BrNO₃
Melting Pt: 112 °C
MDL Number: MFCD06656567
CAS Number: 5847-59-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6ca2b34e5565b637da82fade3fcdd1687ec4b8c&vid=e6ca2b34e5565b637da82fade3fcdd1687ec4b8c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty