Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Chlorobutyric acid ≥90.0%

Supplier: Decon Laboratories Ltd

C0146-25ML C0146-5ML
TCC0146-25MLEA 378 USD InStock
TCC0146-25ML TCC0146-5ML
(±)-2-Chlorobutyric acid ≥90.0%
(±)-2-Chlorobutyric acid
CAS Number: 4170-24-5
MDL Number: MFCD00021697
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 112
Specific Gravity (20/20): 1.19


Formula: C₄H₇ClO₂
MW: 122.55
Boiling Pt: 203
Melting Pt: 37
Density: 1.19 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00021697
CAS Number: 4170-24-5
UN: 3265
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead3a8fa57b0821b84a8fc5e6d0da68449390ecf&vid=ead3a8fa57b0821b84a8fc5e6d0da68449390ecf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty