Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Chloromethyl)trimethylsilane ≥98.0%

Supplier: Decon Laboratories Ltd

C0862-250ML C0862-25ML C0862-100ML
TCC0862-250MLEA 341 USD InStock
TCC0862-250ML TCC0862-25ML TCC0862-100ML
(Chloromethyl)trimethylsilane ≥98.0%
(Chloromethyl)trimethylsilane
CAS Number: 2344-80-1
MDL Number: MFCD00000878
Molecular Formula: C4H11ClSi
Molecular Weight: 122.67
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): -2
Specific Gravity (20/20): 0.88

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Trimethylsilyl)methyl Chloride


Formula: ClH₂CSi(CH₃)₃
Boiling Pt: 98 °C
Density: 0.88 (20 °C)
Flash Pt: –2 °C
MDL Number: MFCD00000878
CAS Number: 2344-80-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4fc314831c53b50d88e5cee5778dd531b2b817&vid=ea4fc314831c53b50d88e5cee5778dd531b2b817&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty