Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexyl vinyl ether ≥98.0% (by GC) stabilized

Supplier: Decon Laboratories Ltd

C0885-25ML C0885-500ML
TCC0885-25MLEA 59.02 USD InStock
TCC0885-25ML TCC0885-500ML
Cyclohexyl vinyl ether ≥98.0% (by GC) stabilized
Cyclohexyl vinyl ether
(stabilized with KOH)
CAS Number: 2182-55-0
MDL Number: MFCD00046358
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 35
Specific Gravity (20/20): 0.89


Formula: C₈H₁₄O
Boiling Pt: 152 °C
Density: 0.89 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00046358
CAS Number: 2182-55-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8bb8fd43d495c988c942314b55043405046c557&vid=f8bb8fd43d495c988c942314b55043405046c557&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty