Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutanecarboxylic acid ≥97.0%

Supplier: Decon Laboratories Ltd

C0888-10G C0888-25G
TCC0888-10GEA 35.59 USD InStock
TCC0888-10G TCC0888-25G
Cyclobutanecarboxylic acid ≥97.0%
Cyclobutanecarboxylic acid
CAS Number: 3721-95-7
MDL Number: MFCD00001323
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 83
Specific Gravity (20/20): 1.06


Formula: C₅H₈O₂
MW: 100.12
Boiling Pt: 196 °C
Melting Pt: -20
Density: 1.06
Flash Pt: 83 °C
MDL Number: MFCD00001323
CAS Number: 3721-95-7
UN: 3265
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=828e15ac90e2750d824be5ce0e620b84f2f684a1&vid=828e15ac90e2750d824be5ce0e620b84f2f684a1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty