Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorocarbonylsulfenyl chloride ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

C1043-25G C1043-5G
TCC1043-25GEA 182 USD InStock
TCC1043-25G TCC1043-5G
Chlorocarbonylsulfenyl chloride ≥97.0% (by titrimetric analysis)
Chlorocarbonylsulfenyl chloride
CAS Number: 2757-23-5
MDL Number: MFCD00000703
Molecular Formula: CCl2OS
Molecular Weight: 130.97
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 62
Specific Gravity (20/20): 1.58

Product Categories: Synthetic Organic Chemistry, Sulfenic Acid Derivatives
Other Product Names / Synonyms: Chloroformylsulfenyl Chloride, Chlorothioformyl Chloride


Formula: CCl₂OS
Boiling Pt: 98 °C
Density: 1.58 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00000703
CAS Number: 2757-23-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9793cda5324a6421d7c669d4a14d555aa97906b8&vid=9793cda5324a6421d7c669d4a14d555aa97906b8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty