Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Chloroacetophenone ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd

C1084-250G C1084-25G
TCC1084-250GEA 218 USD InStock
TCC1084-250G TCC1084-25G
2'-Chloroacetophenone ≥97.0% (by GC)
2'-Chloroacetophenone
CAS Number: 2142-68-9
MDL Number: MFCD00000560
Molecular Formula: C8H7ClO
Molecular Weight: 154.59
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 92
Specific Gravity (20/20): 1.19


Formula: C₈H₇ClO
Boiling Pt: 229 °C
Density: 1.19 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00000560
CAS Number: 2142-68-9
UN: 3416
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5e78d611a84ef16afb0b22fed9ce371e59d6423&vid=f5e78d611a84ef16afb0b22fed9ce371e59d6423&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty