Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

C1089-25G C1089-5G
TCC1089-25GEA 100.4 USD InStock
TCC1089-25G TCC1089-5G
3-Chlorobenzylamine ≥98.0% (by GC, titration analysis)
3-Chlorobenzylamine
CAS Number: 4152-90-3
MDL Number: MFCD00040752
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 98
Specific Gravity (20/20): 1.16


Formula: C₇H₈ClN
Boiling Pt: 226 °C
Density: 1.16 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00040752
CAS Number: 4152-90-3
UN: 2735
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ecb54b55f885a7b1ff50e43e2d20496efa69881&vid=7ecb54b55f885a7b1ff50e43e2d20496efa69881&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty