Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95.0%

Supplier: Decon Laboratories Ltd

C1250-25G C1250-5G
TCC1250-25GEA 274.94 USD InStock
TCC1250-25G TCC1250-5G
2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95.0%
2-Chloro-2-oxo-1,3,2-dioxaphospholane
CAS Number: 6609-64-9
MDL Number: MFCD00043138
Molecular Formula: C2H4ClO3P
Molecular Weight: 142.47
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 110
Specific Gravity (20/20): 1.55
Storage Temperature: <0°C


Formula: C₂H₄ClO₃P
MW: 142.47 g/mol
Boiling Pt: 91 °C
Density: 1.55 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00043138
CAS Number: 6609-64-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b869fbf3e7adb2004b9e3302694dfe5dbf6f6ca6&vid=b869fbf3e7adb2004b9e3302694dfe5dbf6f6ca6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty