Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl cyanoformate ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

C1266-25G C1266-5G
TCC1266-25GEA 99 USD InStock
TCC1266-25G TCC1266-5G
Ethyl cyanoformate ≥98.0% (by GC)
Ethyl cyanoformate
CAS Number: 623-49-4
MDL Number: MFCD00001836
Molecular Formula: C4H5NO2
Molecular Weight: 99.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 24
Specific Gravity (20/20): 1.01

Other Product Names / Synonyms: Cyanoformic Acid Ethyl Ester


Formula: C₄H₅NO₂
Boiling Pt: 116 °C
Density: 1.01 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00001836
CAS Number: 623-49-4
UN: 3275
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=982313e422c73a648891da52ddf6e56cb3b3d38f&vid=982313e422c73a648891da52ddf6e56cb3b3d38f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty