Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R)-(-)-Camphor-10-sulphonyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

C1308-25G C1308-5G
TCC1308-25GEA 162 USD InStock
TCC1308-25G TCC1308-5G
(1R)-(-)-Camphor-10-sulphonyl chloride ≥98.0%
(1R)-(-)-Camphor-10-sulphonyl chloride
CAS Number: 39262-22-1
MDL Number: MFCD00064155
Molecular Formula: C10H15ClO3S
Molecular Weight: 250.74
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 67
Specific rotation [a]20/D: -29.5 deg (C=1, CHCl3)


Formula: C₁₀H₁₅ClO₃S
Melting Pt: 67 °C
MDL Number: MFCD00064155
CAS Number: 39262-22-1
UN: 3261
ADR: 8,I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea57fe2fcfb0234759f3333099c84de4f5aa1b3&vid=bea57fe2fcfb0234759f3333099c84de4f5aa1b3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty