Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-4-cyanoacetophenone ≥96.0%

Supplier: Decon Laboratories Ltd

C1348-25G C1348-5G
TCC1348-25GEA 106.89 USD InStock
TCC1348-25G TCC1348-5G
ω-Bromo-4-cyanoacetophenone ≥96.0%
ω-Bromo-4-cyanoacetophenone
CAS Number: 20099-89-2
MDL Number: MFCD00052931
Molecular Formula: C9H6BrNO
Molecular Weight: 224.06
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 93
Flash Point (°C): 109

Other Product Names / Synonyms: 4-(Bromoacetyl)benzonitrile, 2-Bromo-4'-cyanoacetophenone


Formula: C₉H₆BrNO
Melting Pt: 93 °C
Flash Pt: 109 °C
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2923
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08195a7c89798206ee29e7c4a57be9485a00d251&vid=08195a7c89798206ee29e7c4a57be9485a00d251&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty