Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-(chloromethyl)propene ≥98.0%

Supplier: Decon Laboratories Ltd

C1565-25G C1565-5G
TCC1565-25GEA 256.64 USD InStock
TCC1565-25G TCC1565-5G
3-Chloro-2-(chloromethyl)propene ≥98.0%
3-Chloro-2-(chloromethyl)propene
CAS Number: 1871-57-4
MDL Number: MFCD00000952
Molecular Formula: C4H6Cl2
Molecular Weight: 124.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Melting point (°C): -14
Flash Point (°C): 36
Specific Gravity (20/20): 1.18
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 1,1-Bis(chloromethyl)ethylene, Methallyl Dichloride


Formula: C₄H₆Cl₂
Boiling Pt: 138 °C
Melting Pt: –14 °C
Density: 1.18 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00000952
CAS Number: 1871-57-4
UN: 1992
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4ab6c2421a84ac8d160e31e027f498c588c7695&vid=c4ab6c2421a84ac8d160e31e027f498c588c7695&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty