C1789-25G
C1789-5G
TCC1789-25GEA
108.8
USD
InStock
TCC1789-25G
TCC1789-5G
5-Chlorovaleryl chloride ≥98.0% (by GC, titration analysis)
5-Chlorovaleryl chloride
CAS Number: 1575-61-7
MDL Number: MFCD00000758
Molecular Formula: C5H8Cl2O
Molecular Weight: 155.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 90
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: 5-Chloropentanoyl Chloride