Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroisonicotinonitrile ≥98.0%

Supplier: Decon Laboratories Ltd

C1873-25G C1873-5G
TCC1873-25GEA 105 USD InStock
TCC1873-25G TCC1873-5G
2-Chloroisonicotinonitrile ≥98.0%
2-Chloroisonicotinonitrile
CAS Number: 33252-30-1
MDL Number: MFCD00174318
Molecular Formula: C6H3ClN2
Molecular Weight: 138.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 106
Melting point (°C): 72

Other Product Names / Synonyms: 2-Chloroisonicotinonitrile, 2-Chloro-4-pyridinecarbonitrile


Formula: C₆H₃ClN₂
Boiling Pt: 106 °C
Melting Pt: 72 °C
MDL Number: MFCD00174318
CAS Number: 33252-30-1
UN: 3439
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb7ba552ab70600557ec3125c42ee4d141801700&vid=fb7ba552ab70600557ec3125c42ee4d141801700&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty