Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexyl(dimethoxy)methylsilane ≥98.0%

Supplier: Decon Laboratories Ltd

C1982-25ML C1982-500ML
TCC1982-25MLEA 17 USD InStock
TCC1982-25ML TCC1982-500ML
Cyclohexyl(dimethoxy)methylsilane ≥98.0%
Cyclohexyl(dimethoxy)methylsilane
CAS Number: 17865-32-6
Molecular Formula: C9H20O2Si
Molecular Weight: 188.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 66
Specific Gravity (20/20): 0.94


Formula: C₉H₂₀O₂Si
MW: 188.3394 g/mol
Boiling Pt: 196 °C
Density: 0.94 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00071752
CAS Number: 17865-32-6
UN: 3082
ADR: 9 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20b21a34114426ebb7363aadfe4e5a93bde9e5ae&vid=20b21a34114426ebb7363aadfe4e5a93bde9e5ae&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty