Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane ≥98.0%

Supplier: Decon Laboratories Ltd

C2265-25G C2265-5G
TCC2265-25GEA 171.6 USD InStock
TCC2265-25G TCC2265-5G
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane ≥98.0%
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
CAS Number: 57044-24-3 MDL Number: MFCD00273371 Molecular Formula: C6H11ClO2 Molecular Weight: 150.60 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 63 Flash Point (°C): 51 Specific Gravity (20/20): 1.10 Specific rotation [a]20/D: 44 deg (neat)


Formula: C₆H₁₁ClO₂
Boiling Pt: 63 °C
Density: 1.10 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00273371
CAS Number: 57044-24-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc789532361523efbcfcbb814e9e288c57eb8f3&vid=9dc789532361523efbcfcbb814e9e288c57eb8f3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty