Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methylthiophene ≥96.0%

Supplier: Decon Laboratories Ltd

C2324-25G C2324-5G
TCC2324-25GEA 168.4 USD InStock
TCC2324-25G TCC2324-5G
2-Chloro-5-methylthiophene ≥96.0%
2-Chloro-5-methylthiophene
CAS Number: 17249-82-0
MDL Number: MFCD01632145
Molecular Formula: C5H5ClS
Molecular Weight: 132.61
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.21


Formula: C₅H₅ClS
Boiling Pt: 155 °C
Density: 1.21 (20 °C)
MDL Number: MFCD01632145
CAS Number: 17249-82-0
UN: 1992
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3a5d0f5117c70ca0df44550409afd309f78c158&vid=e3a5d0f5117c70ca0df44550409afd309f78c158&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty