C2391-25G
C2391-5G
TCC2391-25GEA
152
USD
InStock
TCC2391-25G
TCC2391-5G
Cyclopentanecarbonyl chloride ≥98.0% (by GC, titration analysis)
Cyclopentanecarbonyl chloride
CAS Number: 4524-93-0
MDL Number: MFCD00001370
Molecular Formula: C6H9ClO
Molecular Weight: 132.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 60
Specific Gravity (20/20): 1.10