C2536-25G
C2536-5G
TCC2536-25GEA
45.66
USD
InStock
TCC2536-25G
TCC2536-5G
Chloroacetamide ≥98.0% (by GC) for biochemical research
Chloroacetamide
[for Biochemical Research]
CAS Number: 79-07-2
MDL Number: MFCD00008027
Molecular Formula: C2H4ClNO
Molecular Weight: 93.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 120
Flash Point (°C): 170