Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Diaminopentane ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D0108-25ML D0108-5ML
TCD0108-25MLEA 248.76 USD InStock
TCD0108-25ML TCD0108-5ML
1,5-Diaminopentane ≥98.0% (by titrimetric analysis)
1,5-Diaminopentane
CAS Number: 462-94-2
MDL Number: MFCD00008239
Molecular Formula: C5H14N2
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 63
Freezing point (°C): 14
Specific Gravity (20/20): 0.87

Product Categories: Materials Chemistry, Monomers, Bioscience, Other Plant Hormones, Polyamines
Other Product Names / Synonyms: Cadaverine, Pentamethylenediamine, 1,5-Pentanediamine


Formula: C₅H₁₄N₂
Boiling Pt: 180 °C
Density: 0.87 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00008239
CAS Number: 462-94-2
UN: 2735
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a50140ef3bad8cbad7c2e7460170aaa6a14e30ea&vid=a50140ef3bad8cbad7c2e7460170aaa6a14e30ea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty