Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexylcarbodiimide ≥98.0%

Supplier: Decon Laboratories Ltd

D0436-25G D0436-400G
TCD0436-25GEA 32.37 USD InStock
TCD0436-25G TCD0436-400G
Dicyclohexylcarbodiimide ≥98.0%
Dicyclohexylcarbodiimide
CAS Number: 538-75-0
MDL Number: MFCD00011659
Molecular Formula: C13H22N2
Molecular Weight: 206.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 124
Flash Point (°C): 138
Freezing point (°C): 34

Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Nucleic Acid Synthesis, Peptide Synthesis, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: DCC


Formula: C₁₃H₂₂N₂
MW: 206.33 g/mol
Boiling Pt: 124 °C
Flash Pt: 138 °C
MDL Number: MFCD00011659
CAS Number: 538-75-0
UN: 2811
ADR: 6.1 III
Merck Index: 13,03123
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9cd24e4357f23433010804ed0c3aca5444f19ea&vid=f9cd24e4357f23433010804ed0c3aca5444f19ea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty