Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylbenzenesulphonyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

D0683-25G D0683-5G
TCD0683-25GEA 85.8 USD InStock
TCD0683-25G TCD0683-5G
2,5-Dimethylbenzenesulphonyl chloride ≥98.0%
2,5-Dimethylbenzenesulphonyl chloride
CAS Number: 19040-62-1
MDL Number: MFCD00024875
Molecular Formula: C8H9ClO2S
Molecular Weight: 204.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 150
Freezing point (°C): 21

Other Product Names / Synonyms: 2,5-Dimethylbenzenesulfonyl Chloride


Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Boiling Pt: 150 °C
MDL Number: MFCD00024875
CAS Number: 19040-62-1
UN: 3265
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d38108005737920b834e38a0125c55d7103dc96f&vid=d38108005737920b834e38a0125c55d7103dc96f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty