Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,4-Dinitrophenylthio)benzothiazole ≥98.0% (by total nitrogen basis)

Supplier: Decon Laboratories Ltd

D0848-25G D0848-500G
TCD0848-25GEA 28 USD InStock
TCD0848-25G TCD0848-500G
2-(2,4-Dinitrophenylthio)benzothiazole ≥98.0% (by total nitrogen basis)
2-(2,4-Dinitrophenylthio)benzothiazole
CAS Number: 17586-89-9
MDL Number: MFCD00059704
Molecular Formula: C13H7N3O4S2
Molecular Weight: 333.34
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 160

Other Product Names / Synonyms: 2-(2,4-Dinitrophenylmercapto)benzothiazole


Formula: C₁₃H₇N₃O₄S₂
Melting Pt: 160 °C
MDL Number: MFCD00059704
CAS Number: 17586-89-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58574a1d1e4396fc0010179261b3ad03bbcd1018&vid=58574a1d1e4396fc0010179261b3ad03bbcd1018&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty