Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylthiazole ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

D1220-25ML D1220-5ML
TCD1220-25MLEA 87 USD InStock
TCD1220-25ML TCD1220-5ML
2,4-Dimethylthiazole ≥99.0% (by GC)
2,4-Dimethylthiazole
CAS Number: 541-58-2 MDL Number: MFCD00014509 Molecular Formula: C5H7NS Molecular Weight: 113.18 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 146 Flash Point (°C): 43 Specific Gravity (20/20): 1.06


Formula: C₅H₇NS
Boiling Pt: 146 °C
Density: 1.06 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00014509
CAS Number: 541-58-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=942538c4618415ca00364078e36134ae24493e54&vid=942538c4618415ca00364078e36134ae24493e54&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty