Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Diazobicyclo[4.3.0]-5-nonene ≥98.0%

Supplier: Decon Laboratories Ltd

D1313-10ML D1313-250ML D1313-25ML
TCD1313-10MLEA 19 USD InStock
TCD1313-10ML TCD1313-250ML TCD1313-25ML
1,5-Diazobicyclo[4.3.0]-5-nonene ≥98.0%
1,5-Diazobicyclo[4.3.0]-5-nonene
CAS Number: 3001-72-7
MDL Number: MFCD00005554
Molecular Formula: C7H12N2
Molecular Weight: 124.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 98
Specific Gravity (20/20): 1.04

Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: DBN


Formula: C₇H₁₂N₂
Boiling Pt: 108 °C
Density: 1.04 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00005554
CAS Number: 3001-72-7
UN: 3267
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e44ff326e1d9c3f54bd11dcfff5fd87502d242&vid=79e44ff326e1d9c3f54bd11dcfff5fd87502d242&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty