Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-α,α,α-trifluorotoluene ≥98.0%

Supplier: Decon Laboratories Ltd

D1318-25G D1318-5G
TCD1318-25GEA 130.8 USD InStock
TCD1318-25G TCD1318-5G
2,5-Dichloro-α,α,α-trifluorotoluene ≥98.0%
2,5-Dichloro-α,α,α-trifluorotoluene
CAS Number: 320-50-3
MDL Number: MFCD00000608
Molecular Formula: C7H3Cl2F3
Molecular Weight: 215.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 76
Specific Gravity (20/20): 1.49

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,4-Dichloro-2-(trifluoromethyl)benzene, 2,5-Dichloro-alpha,alpha,alpha-trifluorotoluene


Formula: C₇H₃Cl₂F₃
Density: 1.49 (20 °C)
MDL Number: MFCD00000608
CAS Number: 320-50-3
UN: 1760
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68b200ae974072665010801c277fa06e5a73cc54&vid=68b200ae974072665010801c277fa06e5a73cc54&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty