Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Difluoroacetic acid ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D1423-25G D1423-5G
TCD1423-25GEA 143.41 USD InStock
TCD1423-25G TCD1423-5G
Difluoroacetic acid ≥97.0% (by titrimetric analysis)
Difluoroacetic acid
CAS Number: 381-73-7
MDL Number: MFCD00004220
Molecular Formula: C2H2F2O2
Molecular Weight: 96.03
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 135
Melting point (°C): 0
Flash Point (°C): 78
Specific Gravity (20/20): 1.52


Formula: C₂H₂F₂O₂
Boiling Pt: 135 °C
Density: 1.52 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00004220
CAS Number: 381-73-7
UN: 3265
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7cc635bc44758434b6df660a2c472cad4ae80d7&vid=d7cc635bc44758434b6df660a2c472cad4ae80d7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty