Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D1527-25G D1527-5G
TCD1527-25GEA 55 USD InStock
TCD1527-25G TCD1527-5G
2,6-Dimethylpyrazine ≥98.0% (by GC, titration analysis)
2,6-Dimethylpyrazine
CAS Number: 108-50-9
MDL Number: MFCD00006148
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 37
Flash Point (°C): 52


Formula: C₆H₈N₂
Boiling Pt: 155 °C
Melting Pt: 37 °C
Flash Pt: 52 °C
MDL Number: MFCD00006148
CAS Number: 108-50-9
UN: 1325
ADR: 4.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1cfd176bdacf05c701d498c0b7dc51c1ce5ae7f&vid=b1cfd176bdacf05c701d498c0b7dc51c1ce5ae7f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty