Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromothiophene ≥95.0%

Supplier: Decon Laboratories Ltd

D1548-25G D1548-5G
TCD1548-25GEA 98 USD InStock
TCD1548-25G TCD1548-5G
2,3-Dibromothiophene ≥95.0%
2,3-Dibromothiophene
CAS Number: 3140-93-0
MDL Number: MFCD00005418
Molecular Formula: C4H2Br2S
Molecular Weight: 241.93
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 51
Specific Gravity (20/20): 2.19


Formula: C₄H₂Br₂S
Boiling Pt: 219 °C
Density: 2.19 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00005418
CAS Number: 3140-93-0
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07c0ab28263b831e24269d158158a02c7e31fc67&vid=07c0ab28263b831e24269d158158a02c7e31fc67&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty