D1586-25G
D1586-5G
TCD1586-25GEA
261
USD
InStock
TCD1586-25G
TCD1586-5G
1,2:7,8-Diepoxyoctane ≥97.0% (by GC)
1,2:7,8-Diepoxyoctane
CAS Number: 2426-07-5
MDL Number: MFCD00005155
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 98
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2:7,8-Diepoxyoctane