Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloromethylenedimethyliminium chloride ≥97.0%

Supplier: Decon Laboratories Ltd

D1597-25G D1597-5G
TCD1597-25GEA 125.16 USD InStock
TCD1597-25G TCD1597-5G
Dichloromethylenedimethyliminium chloride ≥97.0%
Dichloromethylenedimethyliminium chloride
CAS Number: 33842-02-3
MDL Number: MFCD00011870
Molecular Formula: C3H6Cl3N
Molecular Weight: 162.44
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Other Product Names / Synonyms: Dichloromethylenedimethylammonium Chloride, Phosgene-iminium Chloride


Formula: C₃H₆Cl₃N
MW: 162.44 g/mol
MDL Number: MFCD00011870
CAS Number: 33842-02-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53bffbcef6c13a5edc86d9205020e5648aa7f648&vid=53bffbcef6c13a5edc86d9205020e5648aa7f648&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty