Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S,S'-Dimethyl-N-cyanodithioiminocarbonate ≥98.0%

Supplier: Decon Laboratories Ltd

D1798-250G D1798-25G
TCD1798-250GEA 181 USD InStock
TCD1798-250G TCD1798-25G
S,S'-Dimethyl-N-cyanodithioiminocarbonate ≥98.0%
S,S'-Dimethyl-N-cyanodithioiminocarbonate
CAS Number: 10191-60-3
MDL Number: MFCD00009825
Molecular Formula: C4H6N2S2
Molecular Weight: 146.23
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 110

Other Product Names / Synonyms: Dimethyl N-Cyanodithiocarbonimidate


Formula: C₄H₆N₂S₂
MW: 146.23 g/mol
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD00009825
CAS Number: 10191-60-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c81f0db587c7241349e0c2e6d4732cf33578429&vid=3c81f0db587c7241349e0c2e6d4732cf33578429&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty