Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-1,3-dioxan-5-one ≥97.0%

Supplier: Decon Laboratories Ltd

D2397-1G D2397-5G
TCD2397-1GEA 161.92 USD InStock
TCD2397-1G TCD2397-5G
2,2-Dimethyl-1,3-dioxan-5-one ≥97.0%
2,2-Dimethyl-1,3-dioxan-5-one
CAS Number: 74181-34-3 MDL Number: MFCD00671514 Molecular Formula: C6H10O3 Molecular Weight: 130.14 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 67 Specific Gravity (20/20): 1.09 Storage Temperature: 0-10°C


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 67 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00671514
CAS Number: 74181-34-3
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty