Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

D2791-1G D2791-5G
TCD2791-1GEA 67 USD InStock
TCD2791-1G TCD2791-5G
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran ≥98.0% (by HPLC, total nitrogen)
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran
CAS Number: 28286-88-6
MDL Number: MFCD00181519
Molecular Formula: C10H8N2O
Molecular Weight: 172.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 193

Other Product Names / Synonyms: 2-(2,6-Dimethyl-4H-pyran-4-ylidene)propanedinitrile


Formula: C₁₀H₈N₂O
Melting Pt: 193 °C
MDL Number: MFCD00181519
CAS Number: 28286-88-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d46b0767ae7eb895998cd97de7af636ac7e0fc8&vid=0d46b0767ae7eb895998cd97de7af636ac7e0fc8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty