Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Dimethylamino-2-propyne ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D2794-25ML D2794-5ML
TCD2794-25MLEA 256 USD InStock
TCD2794-25ML TCD2794-5ML
1-Dimethylamino-2-propyne ≥98.0% (by GC, titration analysis)
1-Dimethylamino-2-propyne
CAS Number: 7223-38-3
MDL Number: MFCD00008575
Molecular Formula: C5H9N
Molecular Weight: 83.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): -7
Specific Gravity (20/20): 0.79

Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 3-Dimethylamino-1-propyne


Formula: C₅H₉N
Density: 0.79 (20 °C)
Flash Pt: –7 °C
MDL Number: MFCD00008575
CAS Number: 7223-38-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6704dd461ce787637aed5273b348a7d5510588d9&vid=6704dd461ce787637aed5273b348a7d5510588d9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty