Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dinitro-4-(trifluoromethyl)phenol ≥97.0%

Supplier: Decon Laboratories Ltd

D2880-25G D2880-5G
TCD2880-25GEA 221.8 USD InStock
TCD2880-25G TCD2880-5G
2,6-Dinitro-4-(trifluoromethyl)phenol ≥97.0%
2,6-Dinitro-4-(trifluoromethyl)phenol
CAS Number: 393-77-1
MDL Number: MFCD00194260
Molecular Formula: C7H3F3N2O5
Molecular Weight: 252.11
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Other Product Names / Synonyms: 4-Hydroxy-3,5-dinitrobenzotrifluoride, alpha,alpha,alpha-Trifluoro-2,6-dinitro-p-cresol


Formula: C₇H₃F₃N₂O₅
MDL Number: MFCD00194260
CAS Number: 393-77-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf6ab55d348ddf82fa2c419fc9081ca3decfab0&vid=9cf6ab55d348ddf82fa2c419fc9081ca3decfab0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty