Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dibromo-2-cyanoacetamide ≥98.0% (by total nitrogen basis)

Supplier: Decon Laboratories Ltd

D2902-25G D2902-500G
TCD2902-25GEA 32 USD InStock
TCD2902-25G TCD2902-500G
2,2-Dibromo-2-cyanoacetamide ≥98.0% (by total nitrogen basis)
2,2-Dibromo-2-cyanoacetamide
CAS Number: 10222-01-2
MDL Number: MFCD00129791
Molecular Formula: C3H2Br2N2O
Molecular Weight: 241.87
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 125
Flash Point (°C): 180

Other Product Names / Synonyms: 2-Cyano-2,2-dibromoacetamide


Formula: C₃H₂Br₂N₂O
Melting Pt: 125 °C
Flash Pt: 180 °C
MDL Number: MFCD00129791
CAS Number: 10222-01-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=937c54bd0a1454e6a4476c45c0ff69f5e263f1d0&vid=937c54bd0a1454e6a4476c45c0ff69f5e263f1d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty