Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzenesulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D2990-25G D2990-5G
TCD2990-25GEA 253 USD InStock
TCD2990-25G TCD2990-5G
3,4-Dichlorobenzenesulfonyl chloride ≥98.0% (by titrimetric analysis)
3,4-Dichlorobenzenesulphonyl chloride
CAS Number: 98-31-7
Molecular Formula: C6H3Cl3O2S
Molecular Weight: 245.50
Purity/Analysis Method: >98.0% (T)
Form: Wax
Boiling point (°C): 253
Freezing point (°C): 22


Formula: C₆H₃Cl₃O₂S
Boiling Pt: 253 °C
MDL Number: MFCD00041255
CAS Number: 98-31-7
UN: 3265
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3753862fe8d450298b9df63500bfa145ea0b2924&vid=3753862fe8d450298b9df63500bfa145ea0b2924&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty