Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorothiophenol ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd

D3342-25G D3342-5G
TCD3342-25GEA 851 USD InStock
TCD3342-25G TCD3342-5G
2,4-Difluorothiophenol ≥97.0% (by GC)
2,4-Difluorothiophenol
CAS Number: 1996-44-7
Molecular Formula: C6H4F2S
Molecular Weight: 146.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 49
Specific Gravity (20/20): 1.32

Product Categories: Synthetic Organic Chemistry, Thiols, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorothiophenol


Formula: C₆H₄F₂S
Boiling Pt: 59 °C
Density: 1.32 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00203477
CAS Number: 1996-44-7
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bc8340725a96a8c95a074b65bf2bdbeb553e554&vid=5bc8340725a96a8c95a074b65bf2bdbeb553e554&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty