Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D3503-25G D3503-5G
TCD3503-25GEA 262 USD InStock
TCD3503-25G TCD3503-5G
3,5-Dichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3,5-Dichlorobenzenesulphonyl chloride
CAS Number: 705-21-5
MDL Number: MFCD00051698
Molecular Formula: C6H3Cl3O2S
Molecular Weight: 245.50
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 31


Formula: C₆H₃Cl₃O₂S
MDL Number: MFCD00051698
CAS Number: 705-21-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47000cb1cd173e9227a32252919e89b32a787b03&vid=47000cb1cd173e9227a32252919e89b32a787b03&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty