Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate ≥90.0%

Supplier: Decon Laboratories Ltd

D3883-1G D3883-5G
TCD3883-1GEA 197 USD InStock
TCD3883-1G TCD3883-5G
4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate ≥90.0%
4,5-Dimethoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate
CAS Number: 866252-52-0
Molecular Formula: C12H17F3O5SSi
Molecular Weight: 358.40
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Storage Temperature: <0°C

Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid 4,5-Dimethoxy-2-(trimethylsilyl)phenyl Ester, 4,5-Dimethoxy-2-(trimethylsilyl)phenyl Triflate


Formula: C₁₂H₁₇F₃O₅SSi
MW: 358.40 g/mol
CAS Number: 866252-52-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=435ca4b2cbc6d1233e3f060652b448845ce53866&vid=435ca4b2cbc6d1233e3f060652b448845ce53866&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty