Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl bromomalonate ≥95.0%

Supplier: Decon Laboratories Ltd

D3887-25G D3887-5G
TCD3887-25GEA 126 USD InStock
TCD3887-25G TCD3887-5G
Dimethyl bromomalonate ≥95.0%
Dimethyl bromomalonate
CAS Number: 868-26-8
MDL Number: MFCD00025865
Molecular Formula: C5H7BrO4
Molecular Weight: 211.01
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 110
Specific Gravity (20/20): 1.61

Other Product Names / Synonyms: Bromomalonic Acid Dimethyl Ester


Formula: C₅H₇BrO₄
MW: 211.01
Boiling Pt: 225 °C
Density: 1.61 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00025865
CAS Number: 868-26-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=effdebdc441f8464b02ae901618462033092671e&vid=effdebdc441f8464b02ae901618462033092671e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty