Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobutyraldehyde diethyl acetal ≥98.0%

Supplier: Decon Laboratories Ltd

D3968-25G D3968-5G
TCD3968-25GEA 210 USD InStock
TCD3968-25G TCD3968-5G
4-Aminobutyraldehyde diethyl acetal ≥98.0%
4-Aminobutyraldehyde diethyl acetal
CAS Number: 6346-09-4
MDL Number: MFCD00008227
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 62
Specific Gravity (20/20): 0.91

Other Product Names / Synonyms: 4-Aminobutyraldehyde Diethyl Acetal


Formula: C₈H₁₉NO₂
MW: 161.24 g/mol
Boiling Pt: 196 °C
Density: 0.91 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00008227
CAS Number: 6346-09-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06b8cda24b6495da12dcc6f8cdbd3def3acbd297&vid=06b8cda24b6495da12dcc6f8cdbd3def3acbd297&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty