E0136-1G
E0136-25G
E0136-5G
TCE0136-1GEA
30.04
USD
InStock
TCE0136-1G
TCE0136-25G
TCE0136-5G
N-Ethylmaleimide (NEM) ≥98.0%
N-Ethylmaleimide (NEM)
CAS Number: 128-53-0
MDL Number: MFCD00005509
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 46
Flash Point (°C): 73
Storage Temperature: 0-10°C
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Functional Group Blocking Reagents
Other Product Names / Synonyms: NEM