Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Ethanedisulfonic acid dihydrate ≥95.0%

Supplier: Decon Laboratories Ltd

E0453-25G E0453-5G
TCE0453-25GEA 179 USD InStock
TCE0453-25G TCE0453-5G
1,2-Ethanedisulfonic acid dihydrate ≥95.0%
1,2-Ethanedisulphonic acid dihydrate
CAS Number: 110-04-3 MDL Number: MFCD00069901 Molecular Formula: C2H6O6S2 Molecular Weight: 190.18 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 112


Formula: C₂H₆O₆S₂·2H₂O
Melting Pt: 112 °C
MDL Number: MFCD26793415
CAS Number: 5982-56-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c2d29a69bd0d183c74b94ffb67e7f674b6ec7a1&vid=0c2d29a69bd0d183c74b94ffb67e7f674b6ec7a1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty