Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Oxabicyclo[2.2.1]heptane ≥98.0%

Supplier: Decon Laboratories Ltd

E0508-25ML E0508-5ML
TCE0508-25MLEA 1614.2 USD InStock
TCE0508-25ML TCE0508-5ML
7-Oxabicyclo[2.2.1]heptane ≥98.0%
7-Oxabicyclo[2.2.1]heptane
CAS Number: 279-49-2
MDL Number: MFCD00082342
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 13
Specific Gravity (20/20): 0.98

Other Product Names / Synonyms: 7-Oxabicyclo[2.2.1]heptane


Formula: C₆H₁₀O
MW: 98.15 g/mol
Boiling Pt: 120 °C
Density: 0.98 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00082342
CAS Number: 279-49-2
UN: 3271
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a3221b0af069c789555e1dc9eaddc5af15561b8&vid=8a3221b0af069c789555e1dc9eaddc5af15561b8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty